Order Entry
Germany
ContactUsLinkComponent
 
Sodium chloride ≥99.5%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
 :  OMNI LIFE
 
undefined
Sodium chloride ≥99.5%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
 :  OMNI LIFE
 

 

  • Assay:
    Min. 99.5 %
  • Assay (calculated to the dried substance):
    99.0 - 100.5 %
  • Loss on drying (130°C):
    Max. 0.2 %
  • Water insoluble matter:
    Max. 0.005 %
  • pH (5 %, 25 °C):
    5.0 - 8.0
  • Free acid (as HCl):
    Max. 0.005 %
  • Free alkali (as NaOH):
    Max. 0.005 %
  • Aluminium (Al):
    Max. 0.2 ppm
  • Arsenic (As):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.001 %
  • Calcium (Ca):
    Max. 0.002 %
  • Iron (Fe):
    Max. 1 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 0.0005 %
  • Mg, earth alkali metals (as Ca):
    Max. 100 ppm
  • Heavy metals (as Pb):
    Max. 3 ppm
  • Heavy metals (ICP) ACS:
    Max. 5 ppm
  • Bromide (Br):
    Max. 50 ppm
  • Chlorate, Nitrate (as NO₃):
    Max. 0.003 %
  • Iodide (I):
    Max. 0.001 %
  • Iodide (I) test:
    complying
  • Hexacyanoferrate(II) ([Fe(CN)₆]₄⁻):
    Max. 0.0001 %
  • Ferrocyanides:
    complying
  • Nitrite (NO₂):
    complying
  • Phosphate (PO₄):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 10 ppm
  • Total N:
    Max. 0.001 %
  • Acidity or alkalinity:
    complying
  • Appearance of the solution:
    complying