
Sodium carbonate, anhydrous ≥99.8% (dried basis), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:497-19-8
:
Formula:Na₂CO₃
MDL Number:MFCD00003494
:105.99 g/mol
Merck Index:14,08596
Melting Pt:854 °C
Density:2.53 g/cm³ (20 °C)
EINECS:207-838-8
REACH:01-2119485498-19
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (calculated to the dried substance):Min. 99.8 %
- Loss on drying (300°C):Max. 0.5 %
- Water insoluble matter:Max. 0.01 %
- Silver (Ag):Max. 0.0005 %
- Aluminium (Al):Max. 0.001 %
- Arsenic (As):Max. 0.00005 %
- Barium (Ba):Max. 0.0005 %
- Bismuth (Bi):Max. 0.0005 %
- Calcium (Ca):Max. 0.005 %
- Cadmium (Cd):Max. 0.0005 %
- Cobalt (Co):Max. 0.0005 %
- Chromium (Cr):Max. 0.0005 %
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 5 ppm
- Lithium (Li):Max. 0.0005 %
- Potassium (K):Max. 0.005 %
- Manganese (Mn):Max. 0.0005 %
- Magnesium (Mg):Max. 0.002 %
- Molybdenum (Mo):Max. 0.0005 %
- Nickel (Ni):Max. 0.0005 %
- Lead (Pb):Max. 0.0005 %
- Strontium (Sr):Max. 0.0005 %
- Zinc (Zn):Max. 0.0005 %
- Heavy metals (as Pb):Max. 0.0005 %
- Heavy metals (ICP) ACS:Max. 5 ppm
- Chloride (Cl):Max. 0.001 %
- Total N:Max. 0.001 %
- Total S (as SO₄):Max. 0.003 %
- Phosphate (PO₄):Max. 0.001 %
- Silicate (as SiO₂):Max. 0.002 %
- Residual solvents:complying
Specification Test Results
Frequently Bought Together