n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals
About this item
- ≥95% for general laboratory use.
- CAS Number:111-84-2
- Formula:H₃C(CH₂)₇CH₃
- MDL Number:MFCD00009574
- Molecular Weight:128.26 g/mol
- Boiling Pt:2.9 %(V)
- Melting Pt:–53 °C
- Melting Pt:-53 °C
- Density:0.72 g/cm³ (20 °C)
- Flash Pt:31 °C
- Storage Temperature:Ambient
- EINECS:203-913-4
- UN:1920
- ADR:3,III
3 Options Available Below
Product Details & Documents
- ≥95% for general laboratory use.
- CAS Number:111-84-2
- Formula:H₃C(CH₂)₇CH₃
- MDL Number:MFCD00009574
- Molecular Weight:128.26 g/mol
- Boiling Pt:2.9 %(V)
- Melting Pt:–53 °C
- Melting Pt:-53 °C
- Density:0.72 g/cm³ (20 °C)
- Flash Pt:31 °C
- Storage Temperature:Ambient
- EINECS:203-913-4
- UN:1920
- ADR:3,III
Documents
sds_N29406
SDS
Specifications for Product Family
Specification Test Results
- Explosion Limit:2.9 %
- Identity (IR):complying
- Assay (GC):Min. 98.5 %
- Refractive index (n 20/D):1.403 - 1.407
- Autoignition Temperature:401 °F
- Vapor Density:4.41 (vs air)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



