Order Entry
Germany
ContactUsLinkComponent
Acetic acid ≥99.99% (trace metals basis)
Acetic acid ≥99.99% (trace metals basis)
Certificates
undefined
Acetic acid ≥99.99% (trace metals basis)

Specification Test Results

  • Explosion Limit:
    4 %, 59 °F
  • Identity (IR):
    complying
  • Assay:
    99.5 - 100.5 %
  • Metal trace analysis (ICP):
    Max. 100 ppm
  • Refractive Index:
    n20/D 1.371 (lit.)
  • Vapor Density:
    2.07 (vs air)
  • Autoignition Temperature:
    800 °F

Specification Test Results

Related Information