Calcium chloride dihydrate (max. 0.0001% Al)
Catalog # HONE12064.9025
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
Specification Test Results
- Assay:99.0 - 103.0 %
- pH (5 %, 25 °C):4.5 - 8.5
- Free acid (as HCl):Max. 0.005 %
- Free alkali (as CaO):Max. 0.005 %
- Acidity or alkalinity:complying
- Aluminium (Al):Max. 1 ppm
- Arsenic (As):Max. 2 ppm
- Barium (Ba):complying
- Copper (Cu):Max. 0.001 %
- Iron (Fe):Max. 10 ppm
- Mercury (Hg):Max. 1 ppm
- Potassium (K):Max. 0.01 %
- Magnesium (Mg):Max. 0.05 %
- Sodium (Na):Max. 0.02 %
- Lead (Pb):Max. 2 ppm
- Zinc (Zn):Max. 0.002 %
- Heavy metals (as Pb):Max. 10 ppm
- Fluoride (F):Max. 40 ppm
- Sulfate (SO₄):Max. 200 ppm
- Aluminium, iron and phosphates:complying
- Mg and alkali salts (SO₄):Max. 0.5 %
- Residual solvents:complying
- Appearance of the solution:complying
Specification Test Results
About this item
:
:147.01 g/mol
Merck Index:13,01660
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:176 °C
Density:1.85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-140-8
:GHS07
CAS Number:10035-04-8
Formula:CaCl₂·2H₂O
MDL Number:MFCD00149613