Calcium chloride dihydrate ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:10035-04-8
:
Formula:CaCl₂·2H₂O
MDL Number:MFCD00149613
:147.01 g/mol
Merck Index:13,01660
Boiling Pt:1.900 °C (1.013 hPa)
Melting Pt:765 °C
Storage Temperature:Ambient
EINECS:233-140-8
:GHS07
Specification Test Results
- Assay:99 - 105.0 %
- pH (5 %, 25 °C):4.5 - 8.0
- Water insoluble matter:Max. 0.01 %
- Acidity or alkalinity:complying
- Aluminium (Al):Max. 0.0001 %
- Arsenic (As):Max. 1 ppm
- Barium (Ba):Max. 0.005 %
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 0.005 %
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 0.0005 %
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.005 %
- Sodium (Na):Max. 0.005 %
- Nickel (Ni):Max. 0.005 %
- Lead (Pb):Max. 5 ppm
- Strontium (Sr):Max. 0.01 %
- Zinc (Zn):Max. 0.0005 %
- Mg and alkali salts (SO₄):Max. 0.5 %
- Heavy metals (ICP) ACS:Max. 5 ppm
- Ammonium (NH₄):Max. 0.005 %
- Nitrate (NO₃):Max. 0.002 %
- Sulfate (SO₄):Max. 0.005 %
- Oxidizing substances (as NO₃):Max. 0.003 %
- Appearance of the solution:complying
- Colour of the solution:complying
- Turbidity:Max. 3.5 NTU