
Sodium carbonate monohydrate 99.5-100.5% (dried basis), puriss. Ph. Eur., BP, NF, FCC, E500, extra pure, Fluka™
Catalog # HONE13568.9025
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
Specification Test Results
- Assay (excluding dried substance):99.5 - 100.5 %
- Assay of Na₂CO₃:83.0 - 87.5 %
- Loss on drying (300°C):12.0 - 15.0 %
- Loss on drying (105°C, 4 h):12.0 - 15.0 %
- Arsenic (As):Max. 2 ppm
- Copper (Cu):Max. 0.001 %
- Iron (Fe):Max. 10 ppm
- Mercury (Hg):Max. 1 ppm
- Potassium (K):Max. 0.03 %
- Lead (Pb):Max. 2 ppm
- Zinc (Zn):Max. 0.001 %
- Heavy metals (as Pb):Max. 10 ppm
- Bromide (Br):Max. 0.001 %
- Chloride (Cl):Max. 50 ppm
- Sulfate (SO₄):Max. 100 ppm
- Appearance of the solution:complying
- Alkali hydroxide, -bicarbonates:complying
- Residual solvents:complying
Specification Test Results
About this item
:
:124.00 g/mol
Melting Pt:107.85 °C
Density:2.25 g/cm³ (20 °C)
EINECS:207-838-8
:GHS07
Storage Temperature:Ambient
CAS Number:5968-11-6
Formula:Na₂CO₃·1H₂O
MDL Number:MFCD00149177