

Iodine ≥99.8%, solid ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Melting Pt:113.5 °C
Density:4.930 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-442-4
UN:3495
ADR:8,III
:GHS08
:GHS07
Specification Test Results
- Assay:Min. 99.8 %
- Non-volatile matter:Max. 0.01 %
- Bromine and chlorine (as chloride):Max. 0.005 %
- Form:Solid
- Vapor Pressure:0.31 mmHg ( 25 °C)
- Resistivity:1.3E15 μΩ-cm