Aroclor solutions
Supplier: Restek
About this item
Aroclor standards may exhibit lot-to-lot variation.
Certifications: CRM
24 Options Available Below
Product Details & Documents
Aroclor standards may exhibit lot-to-lot variation.
Certifications: CRM
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- REST32081
- REST32085
- REST32064
- REST32065
- REST32066
- REST32067
- REST32068
- REST32069
- REST32070
- REST32299
- REST32456
- REST32076
- REST32082
- REST32086
- REST32007
- REST32008
- REST32009
- REST32010
- REST32011
- REST32012
- REST32039
- REST32409
- REST32410
- REST32006
- Description:
- Aroclor 1242, 50 mg/kg, transformer oil (PCB free), 1 ml/ampoule
- Aroclor 1254, 50 mg/kg, transformer oil (PCB free), 1 ml/ampoule
- Aroclor 1016, 200 µg/ml, isooctane, 1 ml/ampoule
- Aroclor 1221, 200 µg/ml, isooctane, 1 ml/ampoule
- Aroclor 1232, 200 µg/ml, isooctane, 1 ml/ampoule
- Aroclor 1242, 200 µg/ml, isooctane, 1 ml/ampoule
- Aroclor 1248, 200 µg/ml, isooctane, 1 ml/ampoule
- Aroclor 1254, 200 µg/ml, isooctane, 1 ml/ampoule
- Aroclor 1260, 200 µg/ml, isooctane, 1 ml/ampoule
- Aroclor 1016/1260, 200 mg/kg, isooctane,1 ml/ampoule
- Aroclor 1016/1260, 400 mg/kg, acetone,1 ml/ampoule
- Aroclor 1016, 500 mg/kg, transformer oil (PCB free), 1 ml/ampoule
- Aroclor 1242, 500 mg/kg, transformer oil (PCB free), 1 ml/ampoule
- Aroclor 1254, 500 mg/kg, transformer oil (PCB free), 1 ml/ampoule
- Aroclor 1221, 1000 µg/ml, hexane, 1 ml/ampoule
- Aroclor 1232, 1000 µg/ml, hexane, 1 ml/ampoule
- Aroclor 1242, 1000 µg/ml, hexane, 1 ml/ampoule
- Aroclor 1248, 1000 µg/ml, hexane, 1 ml/ampoule
- Aroclor 1254, 1000 µg/ml, hexane, 1 ml/ampoule
- Aroclor 1260, 1000 µg/ml, hexane, 1 ml/ampoule
- Aroclor 1016/1260, 1000 µg/ml, hexane, 1 ml/ampoule
- Aroclor 1262, 1000 mg/kg hexane,1 ml/ampoule
- Aroclor 1268, 1000 mg/kg hexane,1 ml/ampoule
- Aroclor 1016, 2000 µg/ml, hexane, 1 ml/ampoule
- CAS No.:
- 53469-21-9
- 11097-69-1
- 12674-11-2
- 11104-28-2
- 11141-16-5
- 53469-21-9
- 12672-29-6
- 11097-69-1
- 11096-82-5
- 12674-11-2
- 53469-21-9
- 11097-69-1
- 11104-28-2
- 11141-16-5
- 53469-21-9
- 12672-29-6
- 11097-69-1
- 11096-82-5
- —
- 12674-11-2
- Formula:
- C₁₂H₆Cl₄
- C₁₂H₅Cl₅
- C₁₂H₇Cl₃
- —
- —
- C₁₂H₆Cl₄
- C₁₂H₆Cl₄
- C₁₂H₅Cl₅
- C₁₂H₄Cl₆
- C₁₂H₇Cl₃
- C₁₂H₆Cl₄
- C₁₂H₅Cl₅
- —
- —
- C₁₂H₆Cl₄
- C₁₂H₆Cl₄
- C₁₂H₅Cl₅
- C₁₂H₄Cl₆
- —
- C₁₂H₇Cl₃
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- REST32081Aroclor 1242, 50 mg/kg, transformer oil (PCB free), 1 ml/ampouleAroclor 1242, 50 mg/kg, transformer oil (PCB free), 1 ml/ampoule1 mLLog in for availability1 * 1 mL66,50 €Specifications:
More
Specifications
Cat. No.REST32081DescriptionAroclor 1242, 50 mg/kg, transformer oil (PCB free), 1 ml/ampouleCAS No.53469-21-9FormulaC₁₂H₆Cl₄ - REST32085Aroclor 1254, 50 mg/kg, transformer oil (PCB free), 1 ml/ampouleAroclor 1254, 50 mg/kg, transformer oil (PCB free), 1 ml/ampoule1 mLLog in for availability1 * 1 mL66,50 €Specifications:
More
Specifications
Cat. No.REST32085DescriptionAroclor 1254, 50 mg/kg, transformer oil (PCB free), 1 ml/ampouleCAS No.11097-69-1FormulaC₁₂H₅Cl₅ - REST32064Aroclor 1016, 200 µg/ml, isooctane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32064DescriptionAroclor 1016, 200 µg/ml, isooctane, 1 ml/ampouleCAS No.12674-11-2FormulaC₁₂H₇Cl₃ - REST32065Aroclor 1221, 200 µg/ml, isooctane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32065DescriptionAroclor 1221, 200 µg/ml, isooctane, 1 ml/ampouleCAS No.11104-28-2Formula— - REST32066Aroclor 1232, 200 µg/ml, isooctane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32066DescriptionAroclor 1232, 200 µg/ml, isooctane, 1 ml/ampouleCAS No.11141-16-5Formula— - REST32067Aroclor 1242, 200 µg/ml, isooctane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32067DescriptionAroclor 1242, 200 µg/ml, isooctane, 1 ml/ampouleCAS No.53469-21-9FormulaC₁₂H₆Cl₄ - REST32068Aroclor 1248, 200 µg/ml, isooctane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32068DescriptionAroclor 1248, 200 µg/ml, isooctane, 1 ml/ampouleCAS No.12672-29-6FormulaC₁₂H₆Cl₄ - REST32069Aroclor 1254, 200 µg/ml, isooctane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32069DescriptionAroclor 1254, 200 µg/ml, isooctane, 1 ml/ampouleCAS No.11097-69-1FormulaC₁₂H₅Cl₅ - REST32070Aroclor 1260, 200 µg/ml, isooctane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32070DescriptionAroclor 1260, 200 µg/ml, isooctane, 1 ml/ampouleCAS No.11096-82-5FormulaC₁₂H₄Cl₆ - REST32299Aroclor 1016/1260, 200 mg/kg, isooctane,1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32299DescriptionAroclor 1016/1260, 200 mg/kg, isooctane,1 ml/ampouleCAS No.Formula - REST32456Aroclor 1016/1260, 400 mg/kg, acetone,1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32456DescriptionAroclor 1016/1260, 400 mg/kg, acetone,1 ml/ampouleCAS No.Formula - REST32076Aroclor 1016, 500 mg/kg, transformer oil (PCB free), 1 ml/ampouleAroclor 1016, 500 mg/kg, transformer oil (PCB free), 1 ml/ampoule1 mLLog in for availability1 * 1 mL54,00 €Specifications:
More
Specifications
Cat. No.REST32076DescriptionAroclor 1016, 500 mg/kg, transformer oil (PCB free), 1 ml/ampouleCAS No.12674-11-2FormulaC₁₂H₇Cl₃ - REST32082Aroclor 1242, 500 mg/kg, transformer oil (PCB free), 1 ml/ampouleAroclor 1242, 500 mg/kg, transformer oil (PCB free), 1 ml/ampoule1 mLLog in for availability1 * 1 mL54,00 €Specifications:
More
Specifications
Cat. No.REST32082DescriptionAroclor 1242, 500 mg/kg, transformer oil (PCB free), 1 ml/ampouleCAS No.53469-21-9FormulaC₁₂H₆Cl₄ - REST32086Aroclor 1254, 500 mg/kg, transformer oil (PCB free), 1 ml/ampouleAroclor 1254, 500 mg/kg, transformer oil (PCB free), 1 ml/ampoule1 mLLog in for availability1 * 1 mL56,00 €Specifications:
More
Specifications
Cat. No.REST32086DescriptionAroclor 1254, 500 mg/kg, transformer oil (PCB free), 1 ml/ampouleCAS No.11097-69-1FormulaC₁₂H₅Cl₅ - REST32007Aroclor 1221, 1000 µg/ml, hexane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32007DescriptionAroclor 1221, 1000 µg/ml, hexane, 1 ml/ampouleCAS No.11104-28-2Formula— - REST32008Aroclor 1232, 1000 µg/ml, hexane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32008DescriptionAroclor 1232, 1000 µg/ml, hexane, 1 ml/ampouleCAS No.11141-16-5Formula— - REST32009Aroclor 1242, 1000 µg/ml, hexane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32009DescriptionAroclor 1242, 1000 µg/ml, hexane, 1 ml/ampouleCAS No.53469-21-9FormulaC₁₂H₆Cl₄ - REST32010Aroclor 1248, 1000 µg/ml, hexane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32010DescriptionAroclor 1248, 1000 µg/ml, hexane, 1 ml/ampouleCAS No.12672-29-6FormulaC₁₂H₆Cl₄ - REST32011Aroclor 1254, 1000 µg/ml, hexane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32011DescriptionAroclor 1254, 1000 µg/ml, hexane, 1 ml/ampouleCAS No.11097-69-1FormulaC₁₂H₅Cl₅ - REST32012Aroclor 1260, 1000 µg/ml, hexane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32012DescriptionAroclor 1260, 1000 µg/ml, hexane, 1 ml/ampouleCAS No.11096-82-5FormulaC₁₂H₄Cl₆ - REST32039Aroclor 1016/1260, 1000 µg/ml, hexane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32039DescriptionAroclor 1016/1260, 1000 µg/ml, hexane, 1 ml/ampouleCAS No.—Formula— - REST32409Aroclor 1262, 1000 mg/kg hexane,1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32409DescriptionAroclor 1262, 1000 mg/kg hexane,1 ml/ampouleCAS No.Formula - REST32410Aroclor 1268, 1000 mg/kg hexane,1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32410DescriptionAroclor 1268, 1000 mg/kg hexane,1 ml/ampouleCAS No.Formula - REST32006Aroclor 1016, 2000 µg/ml, hexane, 1 ml/ampouleSpecifications:
More
Specifications
Cat. No.REST32006DescriptionAroclor 1016, 2000 µg/ml, hexane, 1 ml/ampouleCAS No.12674-11-2FormulaC₁₂H₇Cl₃
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



