Order Entry
Germany
ContactUsLinkComponent
Stearyl stearate test solutions
Stearyl stearate test solutions
Supplier:  Restek
Certificates
Stearyl stearate test solutions
Supplier:  Restek

Specifications

  • CAS No.:
    2778-96-3
  • Formula:
    C₃₆H₇₂O₂
  • Description:
    Stearyl stearate

Specifications

Frequently Bought Together