Fuel oil #6 standards
Supplier: Restek
Certificates
About this item
A high-viscosity residual type of heavy fuel oil requiring preheating to 104 to 127 °C. This material is also known as residual fuel oil (RFO) or Bunker C.
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.