Order Entry
Germany
ContactUsLinkComponent
Acetic acid 0.5 mol/L 0.5 N, AVS TITRINORM volumetric solution
Acetic acid 0.5 mol/L 0.5 N, AVS TITRINORM volumetric solution
  85803.290
 :  
undefined
Acetic acid 0.5 mol/L 0.5 N, AVS TITRINORM volumetric solution
  85803.290
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Titer (20°C; real value 0.2 % accuracy):
    0.499 -> 0.501 mol/l

 

 

 :
 :60.05 g/mol
 :Ambient
 :64-19-7
 :H₃CCOOH
 :MFCD00036152