Gas calibration mix
Catalog # REST30014
Supplier: Restek
CAS Number:
Specifications
- Packung:Ampoule
- VE:1 mL
- Bezeichnung:Gas calibration mix (7 components)
- Pk Info:1 ml/ampoule
Specifications
About this item
Gas calibration mix consists of seven components, each with 2000 µg/ml in P&T methanol (1 ml/ampoule). Components include bromomethane, 1,3-butadiene, chlorodifluoromethane, chloromethane, dichlorodifluoromethane, trichlorofluoromethane and vinyl chloride.