Copper(II) oxide ≥97.5%, powder, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1317-38-0
:
Formula:CuO
MDL Number:MFCD00010979
:79.55 g/mol
Merck Index:13,02674
Boiling Pt:2000 °C (1013 hPa)
Melting Pt:1326 °C
Density:4.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-269-1
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Assay:Min. 97.5 %
- Assay (Cu):Min. 78 %
- Insoluble matter:Max. 0.015 %
- Water:Max. 0.2 %
- CO3 (Carbonate):Max. 0.3 %
- Cl (Chloride):Max. 0.03 %
- Fe (Iron):Max. 0.03 %