Order Entry
Germany
Orders LinkContactUsLinkComponent
Hand wash, SURE®

Hand wash, SURE®

   Sustainable Options Available
Supplier:   Diversey
Hand wash, SURE®
Supplier:  Diversey
   Sustainable Options Available
Certificates
Hand wash, SURE®

2 Options Available Below

Product Details & Documents

Gentle and effective hand wash with a 100% natural perfume. Cleans hands leaving them hygienically fresh.

  • Free from parabens and other preservatives.
  • Contains plant-based actives from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product or user
  • 100% natural perfume with a refreshing lemon fragrance
  • Skin compatability tested (Patch test)

SURE® is a comprehensive offering of plant-based, cleaning products designed to deliver superior professional results while being safe for people and kind to the environment.

With environmentally-friendly ingredients from renewable resources, SURE® products meet the daily requirements for kitchens, facilities, and personal care.

Certifications: OECD 301B. Cradle to cradle certification at gold level.

Ordering information: 1.3 L cartridge is suitable for Diversey IntelliCare Dispenser (manual/hybrid).

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Documents

Specifications for Product Family

Specifications

  • Catalog No:
  • 115-2954
  • 115-2955
  • Environmentally Preferable:
  • Safer Human and Environmental Health|Sustainable Materials
  • Environmental certification:
  • OECD 301B
    Cradle to creadle certification at gold level

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • 115-2954
    SURE® Hand Wash, 0,5 L
    SURE® Hand Wash, 0,5 L
    Log in for availability
    Log in for availability
    1 * 6 Fl
    1 * 6 Fl - 41,40 €
    41,40 €
    1 * 6 Fl
       41,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    115-2954
    Environmentally Preferable
    Environmental certification
  • 115-2955
    SURE® Hand Wash, 1,3 L
    SURE® Hand Wash, 1,3 L
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 43,70 €
    43,70 €
    1 * 4 ST
       43,70 €

    Specifications:
      Safer Human and Environmental Health|Sustainable Materials
    More
    Specifications
    Cat. No.
    115-2955
    Environmentally Preferable
    Safer Human and Environmental Health|Sustainable Materials
    Environmental certification
    OECD 301B
    Cradle to creadle certification at gold level
      Safer Human and Environmental Health|Sustainable Materials