Sterlisation flat pouches, Stericlin®
Supplier: VP Medical Packaging
Certificates
About this item
These see-through sterilisation flat pouches made of sterilisation proof 70 g/m² heavy-duty paper are a universal packaging solution.
- Effective barrier against germs and bacteria
- Permits the transmission of air, steam or sterilisation gases
- Three indicators for STEAM, EO and FORM sterilisation are imprinted outside the filling space
Suitable for sterilisation applications of almost all light and medium weight instruments and sets.
Certifications: DIN EN ISO 11607 and DIN 868-5.