Sterilisation gusseted reels, Stericlin®
These see-through sterilisation reels made of sterilisation proof 70 g/m2 heavy-duty paper are a universal packaging solution.
- Effective barrier against germs and bacteria
- Permits the transmission of air, steam or sterilisation gases
- Three indicators for STEAM-, EO- and FORM-sterilisation are imprinted outside the filling space
Suitable for sterilisation applications of almost all light and medium weight instruments and sets.
: DIN EN ISO 11607 and DIN 868-5.