Order Entry
Germany
ContactUsLinkComponent
O-rings for desiccators DURAN®
O-rings for desiccators DURAN®
Catalog # SCOT292156108
Supplier:  DWK Life Sciences
undefined
O-rings for desiccators DURAN®
Catalog # SCOT292156108
Supplier:  DWK Life Sciences

Specifications

  • Ø:
    236 mm
  • For:
    DN 200 desiccators
  • Description:
    O-Ring, silicone (VMQ), DN 200, 236×5,3 mm
  • Seq:
    04
  • Pk:
    1

Specifications

About this item