Order Entry
Germany
ContactUsLinkComponent
Analytical standard, Phenmedipham-(methoxy-d₃), reference material, Honeywell Fluka™
Analytical standard, Phenmedipham-(methoxy-d₃), reference material, Honeywell Fluka™
Catalog # HONCR1302-10MG
CAS Number:  
Analytical standard, Phenmedipham-(methoxy-d₃), reference material, Honeywell Fluka™
Catalog # HONCR1302-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1773497-41-8
  • Description:
    Phenmedipham-(methoxy-d₃)
  • Melting point:
    139…142 °C
  • Formula:
    C₁₆D₃H₁₃N₂O₄
  • Storage temperature:
    Ambient
  • Description:
    Phenmedipham-(methoxy-d₃)

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results