Order Entry
Germany
ContactUsLinkComponent
Analytical standard, Malathion diacid-(dimethyl-d₆), reference material, Honeywell Fluka™
Analytical standard, Malathion diacid-(dimethyl-d₆), reference material, Honeywell Fluka™
  HONCR1279-10MG
 :  
Analytical standard, Malathion diacid-(dimethyl-d₆), reference material, Honeywell Fluka™
  HONCR1279-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1346601-20-4
  • Description:
    Malathion diacid-(dimethyl-d₆)
  • Melting point:
    117…212 °C
  • Formula:
    C₆D₆H₅O₆PS₂
  • Storage temperature:
    2…8 °C
  • Description:
    Malathion diacid-(dimethyl-d₆)

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results