Analytical standard, Prochloraz Metabolite BTS44595, reference material, Honeywell Fluka™
HONCR1277-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:139542-32-8
- Description:Prochloraz Metabolite BTS44595
- Melting point:79…82 °C
- Formula:C₁₃H₁₅Cl₃N₂O₃
- Storage temperature:Ambient
- Description:Prochloraz Metabolite BTS44595
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results