Analytical standard, Ciprofloxacin-d₈ hydrochloride hydrate, reference material, Honeywell Fluka™
HONCR1269-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1216659-54-9
- Description:Ciprofloxacin-d₈ hydrochloride hydrate
- Melting point:min. 310 °C
- Formula:C₁₇D₈H₁₀FN₃O₃ · HCl · xH₂O
- Storage temperature:Ambient
- Description:Ciprofloxacin-d₈ hydrochloride hydrate
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results