Order Entry
Germany
ContactUsLinkComponent
Analytical standard, Albendazole-2-aminosulfone-(propyl-3,3,3-d₃) hydrochloride, reference material, Honeywell Fluka™
Analytical standard, Albendazole-2-aminosulfone-(propyl-3,3,3-d₃) hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1262-10MG
CAS Number:  
Analytical standard, Albendazole-2-aminosulfone-(propyl-3,3,3-d₃) hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1262-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1435902-07-0
  • Description:
    Albendazole-2-aminosulfone-(propyl-3,3,3-d₃) hydrochloride
  • Melting point:
    239…243 °C
  • Formula:
    C₁₀D₃H₁₀N₃O₂S · HCl
  • Storage temperature:
    Ambient
  • Description:
    Albendazole-2-aminosulfone-(propyl-3,3,3-d₃) hydrochloride

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results