Analytical standard, Deterenol acetate, reference material, Honeywell Fluka™
Catalog # HONCR1236-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1644449-83-1
- Description:Deterenol acetate
- Melting point:123…129 °C
- Formula:C₁₁H₁₇NO₂ · C₂H₄O₂
- Storage temperature:Ambient
- Description:Deterenol acetate
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results