Analytical standard, Acetaminophen-(methyl-d₃), reference material, Honeywell Fluka™
Catalog # HONCR1218-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:60902-28-5
- Description:Acetaminophen-(methyl-d₃)
- Melting point:165…170 °C
- Formula:C₈D₃H₆NO₂
- Storage temperature:Ambient
- Description:Acetaminophen-(methyl-d₃)
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results