Analytical standard, Parbendazole, reference material, Honeywell Fluka™
HONCR1196-25MG
:
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:14255-87-9
- Description:Parbendazole
- Melting point:255…257 °C
- Formula:C₁₃H₁₇N₃O₂
- Storage temperature:2…8 °C
- Description:Parbendazole
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results