Analytical standard, Cefoperazone dihydrate, reference material, Honeywell Fluka™
Catalog # HONCR1191-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard, neat
- CAS No.:113826-44-1
- Description:Cefoperazone dihydrate
- Melting point:169…171 °C
- Formula:C₂₅H₂₇N₉O₈S₂ · 2H₂O
- Storage temperature:2…8 °C
- Description:Cefoperazone dihydrate
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results