Analytical standard, Meropenem trihydrate, reference material, Honeywell Fluka™
Catalog # HONCR1186-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:119478-56-7
- Description:Meropenem trihydrate
- Melting point:205 °C
- Formula:C₁₇H₂₅N₃O₅S · 3H₂O
- Storage temperature:Ambient
- Description:Meropenem trihydrate
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results