Analytical standard, Methyl 4-hydroxybenzoate-(ring-¹³C₆), reference material, Honeywell Fluka™
Catalog # HONCR1148-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1581694-95-2
- Description:Methyl 4-hydroxybenzoate-(ring-¹³C₆)
- Melting point:125 °C
- Formula:¹³C₆C₂H₈O₃
- Storage temperature:2…8 °C
- Description:Methyl 4-hydroxybenzoate-(ring-¹³C₆)
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results