Analytical standard, Doripenem monohydrate, reference material, Honeywell Fluka™
Catalog # HONCR1084-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:364622-82-2
- Description:Doripenem monohydrate
- Melting point:185 °C
- Formula:C₁₅H₂₄N₄O₆S₂ · H₂O
- Storage temperature:2…8 °C
- Description:Doripenem monohydrate
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results