Order Entry
Germany
ContactUsLinkComponent
 
Analytical standard, Metalaxyl Metab CGA 62826 , reference material, Honeywell Fluka™
  HONCR1070-25MG
 :  
Analytical standard, Metalaxyl Metab CGA 62826 , reference material, Honeywell Fluka™
  HONCR1070-25MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    25 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    87764-37-2
  • Description:
    Metalaxyl Metab CGA 62826
  • Melting point:
    142…146 °C
  • Formula:
    C₁₄H₁₉NO₄
  • Storage temperature:
    Ambient
  • Description:
    Metalaxyl Metab CGA 62826

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results