Analytical standard, Staurosporine, reference material, Honeywell Fluka™
HONCR1061-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:62996-74-1
- Description:Staurosporine
- Melting point:288…291 °C
- Formula:C₂₈H₂₆N₄O₃
- Storage temperature:–20°C
- Description:Staurosporine
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results