Order Entry
Germany
ContactUsLinkComponent
Analytical standard, Desnitro-imidacloprid hydrochloride, reference material, Honeywell Fluka™
Analytical standard, Desnitro-imidacloprid hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1058-25MG
CAS Number:  
Analytical standard, Desnitro-imidacloprid hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1058-25MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    25 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    127202-53-3
  • Description:
    Desnitro-imidacloprid hydrochloride
  • Melting point:
    250 °C
  • Formula:
    C₉H₁₁ClN₄ · HCl
  • Storage temperature:
    Ambient
  • Description:
    Desnitro-imidacloprid hydrochloride

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results