Analytical standard, 2-(Diethyl-d₁₀-amino)-6-methyl-4-pyrimidinol, reference material, Honeywell Fluka™
Catalog # HONCR1052-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- Description:2-(Diethyl-d₁₀-amino)-6-methyl-4-pyrimidinol
- Melting point:131…137 °C
- Formula:C₉D₁₀H₅N₃O
- Storage temperature:2…8 °C
- Description:2-(Diethyl-d₁₀-amino)-6-methyl-4-pyrimidinol
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results