Order Entry
Germany
ContactUsLinkComponent
 
Analytical standard, Prometryn-(N2-isopropyl-d7) reference material, Honeywell Fluka™
  HONCR1044-10MG
 :  
Analytical standard, Prometryn-(N2-isopropyl-d7) reference material, Honeywell Fluka™
  HONCR1044-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1705649-52-0
  • Description:
    Prometryn-(N2-isopropyl-d7)
  • Melting point:
    104…108 °C
  • Formula:
    C₁₀D₇H₁₂N₅S
  • Storage temperature:
    Ambient
  • Description:
    Prometryn-(N2-isopropyl-d7)

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results