Order Entry
Germany
ContactUsLinkComponent
Analytical standard, Adapalene-(methoxy-d₃), reference material, Honeywell Fluka™
Analytical standard, Adapalene-(methoxy-d₃), reference material, Honeywell Fluka™
  HONCR1038-10MG
 :  
Analytical standard, Adapalene-(methoxy-d₃), reference material, Honeywell Fluka™
  HONCR1038-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1276433-89-6
  • Description:
    Adapalene-(methoxy-d₃)
  • Melting point:
    317…319 °C
  • Formula:
    C₂₈D₃H₂₅O₃
  • Storage temperature:
    Ambient
  • Description:
    Adapalene-(methoxy-d₃)

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results