Analytical standard, Oxfendazole-d₃, reference material, Honeywell Fluka™
Catalog # HONCR1015-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1228182-54-4
- Description:Oxfendazole-d₃
- Melting point:294…299 °C
- Formula:C₁₅D₃H₁₀N₃O₃S
- Storage temperature:Ambient
- Description:Oxfendazole-d₃
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results