Analytical standard, Pirbuterol acetate, 010GFL, reference material, Honeywell Fluka™
HONCR1006-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:65652-44-0
- Description:Pirbuterol acetate, 010GFL
- Melting point:129…132 °C
- Formula:C₁₂H₂₀N₂O₃ · C₂H₄O₂
- Storage temperature:Ambient
- Description:Pirbuterol acetate, 010GFL
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results