Analytical standard, Miloxacin-d₃, reference material, Honeywell Fluka™
HONCR1002-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1228182-43-1
- Description:Miloxacin-d₃
- Melting point:260…265 °C
- Formula:C₁₂D₃H₆NO₆
- Storage temperature:Ambient
- Description:Miloxacin-d₃
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results