Order Entry
Germany
ContactUsLinkComponent
D-rings, DBI-SALA, 3M™
D-rings, DBI-SALA, 3M™
Catalog # CPSD1500003
Supplier:  CAPITAL SAFETY
undefined
D-rings, DBI-SALA, 3M™
Catalog # CPSD1500003
Supplier:  CAPITAL SAFETY

Specifications

  • Description:
    D-ring for 0,9 kg, 13×57 mm
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

D-rings can be attached to virtually any tool under 2,3 kg (5 lbs.) or 0,9 kg (2 lbs.) by using Quick-Wrap Tape.

  • Creates an attachment point in less than a minute
  • Dual D-rings allow for 100% tool tie-off during tool transfers