Body showers
Supplier: BROEN-LAB
Certificates
About this item
Body shower for exposed pipework with self-draining head. In brass with chemical resistant BROEN-LAB Polycoat.
- Professional solutions for employee safety
- Easy to mount
- Anti-limescale design and material
- Improved functionality with long lifetime
- Strong wall flange with 6 holes for wall mounted models with exposed piping
Min. working pressure: 1 bar
Certifications: EN 15154-1
Ordering information: Supporting wall brackets for mounting must be ordered separately.