Order Entry
Germany
ContactUsLinkComponent
VWR® pHenomenal® IDP 761-C Conductivity Cell
VWR® pHenomenal® IDP 761-C Conductivity Cell
Catalog # 663-0356
Supplier:  VWR Collection
undefined
VWR® pHenomenal® IDP 761-C Conductivity Cell
Catalog # 663-0356
Supplier:  VWR Collection

Specifications

  • Conductivity:
    1 µS/cm - 2000 mS/cm
  • Connection:
    IDP plug, 1,5 m cable
    and universal connector
  • Ø×L:
    15,3×120 mm
  • Type:
    pHenomenal® IDP 761-C
  • Description:
    pHenomenal® IDP 761-C intelligent conductivity cell, 4-cell graphite
  • Material:
    Epoxy
  • Pk:
    1
  • Temp.:
    0...+100 °C
  • Cell constant:
    0,475 cm⁻¹

Specifications

About this item

This IDP (Intelligent Digital Probe) conductivity/temperature cell can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter meters.

  • Calibration data (serial number, calibration status) is stored in the electrode/probe, transmitted to the meter and displayed
  • Proven reliability and accuracy
  • Correct measurement results due to direct signal processing in the sensor and digital transmission of values
  • All parameters are easily measured with cable lengths of up to 100 metres
  • Same universal connector for all electrodes and probes

Temperature probe: NTC 30 kOhm

The electrodes can provide the following data, shown on the meter's screen
- Model name
- Serial number
- Calibration status (+++, ++, +, –, blank)
- Calibration history includes the last 10 calibrations, including buffers used, mV values, etc.

Delivery: Includes digital 4-electrode conductivity cell and manual only.