Order Entry
Germany
ContactUsLinkComponent
Alcometer fuel cell breathalyser
Alcometer fuel cell breathalyser
Catalog # SUREALCOM
Supplier:  SURESCREEN
Alcometer fuel cell breathalyser
Catalog # SUREALCOM
Supplier:  SURESCREEN

Specifications

  • Range:
    0,00 - 2,00 mg/l
  • Description:
    Fuell cell Alcometer
  • Accuracy:
    ±0,005% BAC at 0,050% BAC
  • Pk:
    1 KIT

Specifications

About this item

The Alcometer is a fuel cell breathalyser offering reliable, accurate results using the gold standard in breath alcohol testing.

  • High accuracy and long life
  • Flow sensor to indicate donor is using the device correctly
  • Backlit displays for use at night
  • Increased testing speed
  • Warning lights for positive results
  • 6 months/500 uses between calibrations

Results in BrAC.

Delivery: Kit comes with 3 mouthpieces, battery and a heavy duty case.