Order Entry
Germany
ContactUsLinkComponent
Calbiochem® Human Transferrin (apo) (from plasma)
Calbiochem® Human Transferrin (apo) (from plasma)
  616395-100
 :  
Calbiochem® Human Transferrin (apo) (from plasma)
  616395-100
 :  

 

  • Pack type:
    Plastic ampoule
  • Pk:
    100 mg
  • CAS No.:
    11096-37-0
  • Source:
    Plasma
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Form:
    Lyophilised solid
  • Protein/peptide name:
    Transferrin (apo)
  • Purity:
    >95%
  • Formulation:
    Lyophilised from 10 mM sodium phosphate buffer, pH 7,4

 

 

Native apo-transferrin from human plasma.

  • Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV

Transferrin is a serum protein involved in iron binding and transport. Substantially iron-free.