ContactUsLinkComponent
Column cutter
Column cutter
Catalog # PERCN9301376
Supplier:  PerkinElmer
Column cutter
Catalog # PERCN9301376
Supplier:  PerkinElmer
Specifications
  • Description:
    Wafer scribe
  • Pk:
    10
Specifications
About this item
Ceramic wafer scribe is inexpensive and ideal for cutting polyimide fused silica capillary columns and guard columns.
  • Easy to hold
  • All four sides can be used as a cutting tool