Order Entry
Germany
ContactUsLinkComponent
Prostaglandin D₂ Express ELISA kit
Prostaglandin D₂ Express ELISA kit
Supplier:  Cayman Chemical
Certificates
undefined
Prostaglandin D₂ Express ELISA kit
Supplier:  Cayman Chemical

Specifications

  • Assay Type:
    Competitive
  • Target protein:
    PGD₂
  • Developmental time:
    60 min
  • Mid point:
    800 - 1500 pg/ml (50% B/B0)
  • Detection range:
    117,2 - 15000 pg/ml
  • Storage temperature:
    −80 °C
  • Sensitivity:
    350 pg/ml (80% B/B0)
  • Incubation time:
    2 hr

Specifications