Order Entry
Germany
Orders LinkContactUsLinkComponent
Carbonate buffer concentrate

Carbonate buffer concentrate

Carbonate buffer concentrate, 1 L
Catalog # YSIL1579
Supplier:   YSI LIFE SCIENCE
Carbonate buffer concentrate
Catalog # YSIL1579
Supplier:  YSI LIFE SCIENCE
CAS Number:  
Carbonate buffer concentrate

Carbonate buffer concentrate, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents

Carbonate buffer concentrate for use with methanol membranes and ethanol membranes. Reconstitute to make 1 litre.

  • Equipment used with: 2900, 7100, 2700, 2950

Caution: Store at 4 to 25 °C

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Carbonate buffer concentrate

Specifications