Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer solution pH 1,09

Buffer solution pH 1,09

Supplier:   Bernd Kraft
Buffer solution pH 1,09
Supplier:  Bernd Kraft
Certificates
Buffer solution pH 1,09

7 Options Available Below

Product Details & Documents
  • Stabilized

Certifications: According to DIN 19267.

Specifications for Product Family

Specifications

Product Family Options

Product Information

  • pH value
    Pk
    Availability
    Price
  • KRAF03853.2000
    Buffer solution
    Buffer solution
    1,09 (25 °C)
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 39,80 €
    39,80 €
    1 * 500 mL
       39,80 €

    Specifications:
    More
    Specifications
    Cat. No.
    KRAF03853.2000
  • KRAF03853.3000
    Buffer solution
    Buffer solution
    1,09 (25 °C)
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 60,00 €
    60,00 €
    1 * 1 L
       60,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    KRAF03853.3000
  • KRAF03853.3060
    Buffer solution
    Buffer solution
    1,09 (25 °C)
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 64,40 €
    64,40 €
    1 * 1 L
       64,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    KRAF03853.3060
  • KRAF03853.5000
    Buffer solution
    Buffer solution
    1,09 (25 °C)
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 216,00 €
    216,00 €
    1 * 5 L
       216,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    KRAF03853.5000
  • KRAF03853.5400
    Buffer solution
    Buffer solution
    1,09 (25 °C)
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 221,00 €
    221,00 €
    1 * 5 L
       221,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    KRAF03853.5400
  • KRAF03853.6000
    Buffer solution
    Buffer solution
    1,09 (25 °C)
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 294,00 €
    294,00 €
    1 * 10 L
       294,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    KRAF03853.6000
  • KRAF03853.6400
    Buffer solution
    Buffer solution
    1,09 (25 °C)
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 303,00 €
    303,00 €
    1 * 10 L
       303,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    KRAF03853.6400