Laboratory bottles, pressure resistant, glass
New Product
These bottles are made from pressure resistant borosilicate glass 3.3.
- Without cap
- Without spout
Pressure resistant from –1 to 1,5 bar
: ISO 4796.
These bottles are made from pressure resistant borosilicate glass 3.3.
Pressure resistant from –1 to 1,5 bar