TLC plates, silica gel 60, unmodified layers, SIL HD
Supplier: MACHEREY-NAGEL
Certificates
About this item
Unmodified standard silica layers with "High Definition" properties for TLC.
- High-quality silica gel
- Optimised binder system
- High luminosity and brilliant staining properties
SIL HD glass plates are characterised by good wettability for precise colorisation results, even with 100% aqueous detection reagents. High suitability for trace analyses resulting from a UV indicator with increased brilliance and a low-noise background of the layer.
Silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 – 17 μm.